COMH4211ADEB Combi-Blocks, Inc.

Heptanoic acid, 7-[[(1,1-dimethylethoxy)carbonyl]amino]-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD31381694
  • Cas Number : 441349-04-8
  • Molecular Weight : 342.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-dioxopyrrolidin-1-yl 7-((tert-butoxycarbonyl)amino)heptanoate
Inchi Code 1S/C16H26N2O6/c1-16(2,3)23-15(22)17-11-7-5-4-6-8-14(21)24-18-12(19)9-10-13(18)20/h4-11H2,1-3H3,(H,17,22)
Inchi Key ZFUOZXGXWKTKGY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2392

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2392
Availability details for
Quantity
  • Availability
  • Add to Cart