COMH4211ADEC Combi-Blocks, Inc.

N-(3-Iodo-4-methylphenyl)-3-thiophenecarboxamide

  • MDL number MFCD28128272
  • Cas Number : 623907-55-1
  • Molecular Weight : 343.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2393

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2393
Availability details for
Quantity
  • Availability
  • Add to Cart