COMH4211ADED Combi-Blocks, Inc.

4-Nitro-3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31614008
  • Molecular Weight : 228.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2394
Availability details for
Quantity
  • Availability
  • Add to Cart