COMH4211ADEE Combi-Blocks, Inc.

4-(Heptyloxy)benzohydrazide

  • MDL number MFCD03014576
  • Cas Number : 64328-64-9
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2397

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2397
Availability details for
Quantity
  • Availability
  • Add to Cart