COMH4211ADEF Combi-Blocks, Inc.

(4,4-Difluoro-piperidin-1-yl)-(4-iodo-phenyl)-methanone

  • MDL number MFCD31381587
  • Cas Number : 1228778-07-1
  • Molecular Weight : 351.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2399

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2399
Availability details for
Quantity
  • Availability
  • Add to Cart