COMH4211ADF2 Combi-Blocks, Inc.

4-Bromo-3-fluoro-n-(propan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD31539560
  • Cas Number : 1055995-81-7
  • Molecular Weight : 296.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2402

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2402
Availability details for
Quantity
  • Availability
  • Add to Cart