COMH4211ADF3 Combi-Blocks, Inc.

1-[2-(3-Bromophenyl)ethyl]pyrrolidine

  • MDL number MFCD29957406
  • Cas Number : 1225280-80-7
  • Molecular Weight : 254.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-bromophenethyl)pyrrolidine
Inchi Code 1S/C12H16BrN/c13-12-5-3-4-11(10-12)6-9-14-7-1-2-8-14/h3-5,10H,1-2,6-9H2
Inchi Key HPWUBDIFJUPQKU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2403
Availability details for
Quantity
  • Availability
  • Add to Cart