COMH4211ADF5 Combi-Blocks, Inc.

4-(((4-Methylbenzyl)oxy)methyl)piperidine hydrochloride

  • MDL number MFCD21098450
  • Cas Number : 1353989-66-8
  • Molecular Weight : 255.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2405

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2405
Availability details for
Quantity
  • Availability
  • Add to Cart