COMH4211ADF6 Combi-Blocks, Inc.

2'-Fluoro-4'-isopropoxyacetophenone

  • MDL number MFCD00142714
  • Cas Number : 289039-80-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2407
Availability details for
Quantity
  • Availability
  • Add to Cart