COMH4211ADF7 Combi-Blocks, Inc.

2-(4-Cyclopropylmethoxy-pyrazol-1-yl)-phenylamine

  • MDL number MFCD31579542
  • Cas Number : 2270905-62-7
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2410
Availability details for
Quantity
  • Availability
  • Add to Cart