COMH4211ADF8 Combi-Blocks, Inc.

N-Benzyl-n-hexylamine

  • MDL number MFCD00957890
  • Cas Number : 25468-44-4
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2414
Availability details for
Quantity
  • Availability
  • Add to Cart