COMH4211ADF9 Combi-Blocks, Inc.

5-(4-Bromophenyl)-3-trifluoromethyl-1h-pyrazole

  • MDL number MFCD01554560
  • Molecular Weight : 291.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2416

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2416
Availability details for
Quantity
  • Availability
  • Add to Cart