COMH4211AE4A Combi-Blocks, Inc.

2-tert-Butoxycarbonylamino-3-piperidin-4-yl-propionic acid ethyl ester

  • MDL number MFCD31561581
  • Cas Number : 2103520-09-6
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2421
Availability details for
Quantity
  • Availability
  • Add to Cart