COMH4211AE4D Combi-Blocks, Inc.

1-Ethylpiperidine-4-carbonitrile

  • MDL number MFCD11641477
  • Cas Number : 88654-16-4
  • Molecular Weight : 138.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2430

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2430
Availability details for
Quantity
  • Availability
  • Add to Cart