COMH4211AE4F Combi-Blocks, Inc.

N-(2-Benzoylphenyl)-2-pyridinecarboxamide

  • MDL number MFCD00756959
  • Cas Number : 124522-52-7
  • Molecular Weight : 302.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2436

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2436
Availability details for
Quantity
  • Availability
  • Add to Cart