COMH4211AE50 Combi-Blocks, Inc.

4-(4-Carboxy-3-methyl-phenyl)-3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583149
  • Cas Number : 887971-35-9
  • Molecular Weight : 317.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2437
Availability details for
Quantity
  • Availability
  • Add to Cart