COMH4211AE9C Combi-Blocks, Inc.

3-[2-Amino-3-(2-ethoxycarbonyl-ethoxy)-2-(2-ethoxycarbonyl-ethoxymethyl)-propoxy]-propionic acid ethyl ester

  • MDL number MFCD30145489
  • Cas Number : 132491-90-8
  • Molecular Weight : 421.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2441

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2441
Availability details for
Quantity
  • Availability
  • Add to Cart