COMH4211AE9E Combi-Blocks, Inc.

1-(2-Ethoxy-3-fluoro-phenyl)-propylamine hydrochloride

  • MDL number MFCD31557788
  • Cas Number : 2204052-16-2
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2444

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2444
Availability details for
Quantity
  • Availability
  • Add to Cart