COMH4211AEA0 Combi-Blocks, Inc.

6-(Cyclopentylsulfonyl)nicotinic acid

  • MDL number MFCD31579481
  • Cas Number : 1072855-65-2
  • Molecular Weight : 255.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2448

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2448
Availability details for
Quantity
  • Availability
  • Add to Cart