COMH4211AEA1 Combi-Blocks, Inc.

5-Bromo-2-cyclopropylmethoxy-benzoic acid methyl ester

  • MDL number MFCD31381722
  • Cas Number : 2112645-86-8
  • Molecular Weight : 285.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-(cyclopropylmethoxy)benzoate
Inchi Code 1S/C12H13BrO3/c1-15-12(14)10-6-9(13)4-5-11(10)16-7-8-2-3-8/h4-6,8H,2-3,7H2,1H3
Inchi Key NKQKRVMZTJGVSQ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2449

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2449
Availability details for
Quantity
  • Availability
  • Add to Cart