COMH4211AEA2 Combi-Blocks, Inc.

5-Bromo-n-ethyl-2-(2-methoxy-ethoxy)-benzamide

  • MDL number MFCD07627517
  • Cas Number : 904877-40-3
  • Molecular Weight : 302.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-N-ethyl-2-(2-methoxyethoxy)benzamide
Inchi Code 1S/C12H16BrNO3/c1-3-14-12(15)10-8-9(13)4-5-11(10)17-7-6-16-2/h4-5,8H,3,6-7H2,1-2H3,(H,14,15)
Inchi Key QOLSSCCUHDMHPZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2452
Availability details for
Quantity
  • Availability
  • Add to Cart