COMH4211AEA3 Combi-Blocks, Inc.

2-Fluoro-4-methylbenzylamine hydrochloride

  • MDL number MFCD13194346
  • Cas Number : 1214334-14-1
  • Molecular Weight : 175.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2455
Availability details for
Quantity
  • Availability
  • Add to Cart