COMH4211AEA4 Combi-Blocks, Inc.

2-Ethoxy-5-hydroxy-n,n-dimethyl-benzamide

  • MDL number MFCD31618906
  • Cas Number : 2270908-06-8
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2462

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2462
Availability details for
Quantity
  • Availability
  • Add to Cart