COMH4211AEA5 Combi-Blocks, Inc.

2-(4-Bromo-2-fluoro-phenyl)-n,n-dimethyl-acetamide

  • MDL number MFCD31613953
  • Cas Number : 1788546-51-9
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2464
Availability details for
Quantity
  • Availability
  • Add to Cart