COMH4211AEA7 Combi-Blocks, Inc.

C-(2-Ethoxy-3-methyl-phenyl)-c-phenyl-methylamine hydrochloride

  • MDL number MFCD31557897
  • Cas Number : 2203940-85-4
  • Molecular Weight : 277.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2469

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2469
Availability details for
Quantity
  • Availability
  • Add to Cart