COMH4211AEA8 Combi-Blocks, Inc.

Desloratadine n-hydroxypiperidine

  • MDL number MFCD28898432
  • Cas Number : 1193725-73-3
  • Molecular Weight : 326.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2472
Availability details for
Quantity
  • Availability
  • Add to Cart