COMH4211AEAA Combi-Blocks, Inc.

4-Bromo-1-isobutylpyridin-2(1h)-one

  • MDL number MFCD29045247
  • Cas Number : 1620698-93-2
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2477
Availability details for
Quantity
  • Availability
  • Add to Cart