COMH4211AEAC Combi-Blocks, Inc.

tert-Butyl 4-(4-amino-3-chlorophenyl)piperazine-1-carboxylate

  • MDL number MFCD24467609
  • Cas Number : 1386950-73-7
  • Molecular Weight : 311.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-(4-amino-3-chlorophenyl)piperazine-1-carboxylate
Inchi Code 1S/C15H22ClN3O2/c1-15(2,3)21-14(20)19-8-6-18(7-9-19)11-4-5-13(17)12(16)10-11/h4-5,10H,6-9,17H2,1-3H3
Inchi Key DAHHGOLPHPTCFF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2480
Availability details for
Quantity
  • Availability
  • Add to Cart