COMH4211AEAD Combi-Blocks, Inc.

Methyl 3-[(benzylcarbamoyl)amino]propanoate

  • MDL number MFCD11868550
  • Cas Number : 1146299-57-1
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-{[(benzylamino)carbonyl]amino}propanoate
Inchi Code 1S/C12H16N2O3/c1-17-11(15)7-8-13-12(16)14-9-10-5-3-2-4-6-10/h2-6H,7-9H2,1H3,(H2,13,14,16)
Inchi Key KSHHSVVGOPGGRU-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2481

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2481
Availability details for
Quantity
  • Availability
  • Add to Cart