COMH4211AEAE Combi-Blocks, Inc.

Cyclopropyl-(5-methoxy-2-nitro-benzyl)-amine

  • MDL number MFCD31614076
  • Cas Number : 2270907-47-4
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(5-methoxy-2-nitrobenzyl)cyclopropanamine
Inchi Code 1S/C11H14N2O3/c1-16-10-4-5-11(13(14)15)8(6-10)7-12-9-2-3-9/h4-6,9,12H,2-3,7H2,1H3
Inchi Key HJTHFRNFKPZQFM-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2482
Availability details for
Quantity
  • Availability
  • Add to Cart