COMH4211AEAF Combi-Blocks, Inc.

2-(Pyrrolidin-1-yl)-n-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide

  • MDL number MFCD28805775
  • Cas Number : 825630-80-6
  • Molecular Weight : 330.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2484
Availability details for
Quantity
  • Availability
  • Add to Cart