COMH4211AEB0 Combi-Blocks, Inc.

1-Pyrrolidinehexanoic acid hydrochloride

  • MDL number MFCD29763616
  • Cas Number : 2365419-06-1
  • Molecular Weight : 221.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2487
Availability details for
Quantity
  • Availability
  • Add to Cart