COMH4211AEB2 Combi-Blocks, Inc.

(2-Ethoxy-5-hydroxy-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD31618907
  • Cas Number : 2270907-80-5
  • Molecular Weight : 235.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2495

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2495
Availability details for
Quantity
  • Availability
  • Add to Cart