COMH4211AEB5 Combi-Blocks, Inc.

4-(4,4-Dimethyl-piperidin-1-yl)-butyric acid hydrochloride

  • MDL number MFCD31579336
  • Cas Number : 2270906-34-6
  • Molecular Weight : 235.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2499
Availability details for
Quantity
  • Availability
  • Add to Cart