COMH4211AEB6 Combi-Blocks, Inc.

5-Methyl-2-(pyridin-4-yl)benzoic acid hydrochloride

  • MDL number MFCD31583368
  • Cas Number : 1429505-70-3
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2500

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2500
Availability details for
Quantity
  • Availability
  • Add to Cart