COMH4211AEB8 Combi-Blocks, Inc.

3-[2-(Tetrahydro-pyran-4-yl)-ethyl]-benzoic acid

  • MDL number MFCD31614026
  • Cas Number : 2270912-56-4
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2502

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2502
Availability details for
Quantity
  • Availability
  • Add to Cart