COMH4211AEBA Combi-Blocks, Inc.

3-(Aminomethyl)-n-(2-methylpropyl)benzamide

  • MDL number MFCD09049727
  • Cas Number : 926257-70-7
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2512

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2512
Availability details for
Quantity
  • Availability
  • Add to Cart