COMH4211AEBB Combi-Blocks, Inc.

1-(2,3,6-Trichlorophenyl)ethan-1-amine

  • MDL number MFCD24523297
  • Cas Number : 1337243-83-0
  • Molecular Weight : 224.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2513
Availability details for
Quantity
  • Availability
  • Add to Cart