COMH4211AEBE Combi-Blocks, Inc.

3-(2,2,2-Trifluoro-ethanesulfonyl)-phenylamine

  • MDL number MFCD24062682
  • Cas Number : 1516948-11-0
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2517

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2517
Availability details for
Quantity
  • Availability
  • Add to Cart