COMH4211AEBF Combi-Blocks, Inc.

5-Hydroxy-2-(propan-2-yloxy)benzoic acid

  • MDL number MFCD16997388
  • Cas Number : 1243364-88-6
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2518

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2518
Availability details for
Quantity
  • Availability
  • Add to Cart