COMH4211AEC0 Combi-Blocks, Inc.

N-Cyclobutyl-3-ethylaniline

  • MDL number MFCD16107984
  • Cas Number : 1251074-78-8
  • Molecular Weight : 175.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2520

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2520
Availability details for
Quantity
  • Availability
  • Add to Cart