COMH4211AEC1 Combi-Blocks, Inc.

(R,E)-3-But-2-enoyl-4-phenyloxazolidin-2-one

  • MDL number MFCD28393169
  • Cas Number : 148766-15-8
  • Molecular Weight : 231.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2521

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2521
Availability details for
Quantity
  • Availability
  • Add to Cart