COMH4211AEC6 Combi-Blocks, Inc.

Cyclopropanesulfonic acid (3-bromo-5-methyl-phenyl)-amide

  • MDL number MFCD31557876
  • Cas Number : 1801166-69-7
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2537

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2537
Availability details for
Quantity
  • Availability
  • Add to Cart