COMH4211AEC8 Combi-Blocks, Inc.

3-(3-Phenyl-propylsulfanyl)-propionic acid

  • MDL number MFCD09929811
  • Cas Number : 1016738-65-0
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2544
Availability details for
Quantity
  • Availability
  • Add to Cart