COMH4211AEC9 Combi-Blocks, Inc.

1-Benzyl-4-(6-bromopyridine-2-carbonyl)piperazine

  • MDL number MFCD31628830
  • Cas Number : 1876755-44-0
  • Molecular Weight : 360.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2545
Availability details for
Quantity
  • Availability
  • Add to Cart