COMH4211AECB Combi-Blocks, Inc.

(Benzyloxy)(heptyl)amine

  • MDL number MFCD21116647
  • Cas Number : 112151-61-8
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2547
Availability details for
Quantity
  • Availability
  • Add to Cart