COMH4211AECC Combi-Blocks, Inc.

1-[3-Bromo-5-(trifluoromethyl)phenyl]ethan-1-amine

  • MDL number MFCD18710146
  • Cas Number : 1270417-52-1
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2548

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2548
Availability details for
Quantity
  • Availability
  • Add to Cart