COMH4211AECD Combi-Blocks, Inc.

5-Bromo-2-(2,2,2-trifluoro-ethoxy)-benzylamine

  • MDL number MFCD11195048
  • Cas Number : 1039920-55-2
  • Molecular Weight : 284.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2550

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2550
Availability details for
Quantity
  • Availability
  • Add to Cart