COMH4211AECE Combi-Blocks, Inc.

2,2,2-Trifluoro-n-phenethylethanamine

  • MDL number MFCD09938419
  • Cas Number : 106241-19-4
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2553

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2553
Availability details for
Quantity
  • Availability
  • Add to Cart