COMH4211AECF Combi-Blocks, Inc.

2-(4-(4-Methoxyphenyl)cyclohexyl)acetic acid

  • MDL number MFCD21896095
  • Cas Number : 1368684-36-9
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2555

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-2555
Availability details for
Quantity
  • Availability
  • Add to Cart